Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation Symmetry C18 Column

Catalog No. 50819168 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
40 mm
Particle Size:
3.5 μm
5 μm
7 μm
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
50-819-168 50 mm 4.6 mm 3.5 μm
50-819-167 50 mm 2.1 mm 3.5 μm
50-818-728 150 mm 4.6 mm 3.5 μm
50-819-096 150 mm 2.1 mm 3.5 μm
50-819-043 50 mm 4.6 mm 5 μm
50-780-461 150 mm 3 mm 3.5 μm
50-780-462 100 mm 3 mm 3.5 μm
50-781-656 100 mm 40 mm 7 μm
50-780-114 100 mm 2.1 mm 3.5 μm
50-780-115 75 mm 4.6 mm 3.5 μm
50-780-116 100 mm 4.6 mm 3.5 μm
50-780-125 50 mm 2.1 mm 5 μm
50-780-159 30 mm 4.6 mm 3.5 μm
50-780-456 250 mm 3 mm 5 μm
50-781-238 100 mm 2.1 mm 5 μm
50-781-241 50 mm 3 mm 3.5 μm
50-781-245 100 mm 4.6 mm 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 50-819-168 Supplier Waters Corporation Supplier No. 186000201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica.

  • Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
  • Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.
  • No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
  • Available in a variety of particle sizes, chemistries and dimensions.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 50 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 300 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.