Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation Symmetry C4 Column

Catalog No. 50819169 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
75 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3.9 mm
4.6 mm
Particle Size:
3.5 μm
5 μm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
50-819-169 100 mm 2.1 mm 3.5 μm
50-819-097 150 mm 2.1 mm 3.5 μm
50-818-730 150 mm 4.6 mm 3.5 μm
50-818-731 150 mm 4.6 mm 5 μm
50-780-176 250 mm 4.6 mm 5 μm
50-780-164 50 mm 2.1 mm 3.5 μm
50-780-167 50 mm 4.6 mm 3.5 μm
50-780-168 75 mm 4.6 mm 3.5 μm
50-780-169 100 mm 4.6 mm 3.5 μm
50-780-172 150 mm 2.1 mm 5 μm
50-780-173 150 mm 3.9 mm 5 μm
50-780-174 50 mm 4.6 mm 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 50-819-169 Supplier Waters Corporation Supplier No. 186000278
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.

  • No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
  • Available in a variety of particle sizes, chemistries and dimensions.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 300 Å
Quantity 1/Ea.
USP Type L26
Endcapped Yes
For Use With (Application) Protein
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.