Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYMPK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583614 Supplier Invitrogen™ Supplier No. PA583614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKSRVISELQ EACWDMVSAM AGDIILLLDS DNDGIRTHAI KFVEGLIVTL SPRMADSEIP RRQEHDISLD RIPRDHPYIQ YNVLWE Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 95%.

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYMPK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sympk
Gene Accession No. Q92797
Gene Alias 1500016F02Rik; 4632415H16Rik; AA125406; AI449890; FLJ27092; hepatocyte nuclear factor 3 gamma; Hfn3g; LOW QUALITY PROTEIN: symplekin; SPK; SYM; SYMPK; Symplekin
Gene Symbols Sympk
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYMPK. Recombinant protein control fragment (Product #RP-104754).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.