Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Symplekin Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074590F
Description
Symplekin Polyclonal antibody specifically detects Symplekin in Human samples. It is validated for ImmunohistochemistrySpecifications
Symplekin | |
Polyclonal | |
Immunohistochemistry | |
FLJ27092, SPKSYM, symplekin | |
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Symplekin using the numbering given in entry NP_004810.2 (GeneID 8189). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8189 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction