Learn More
Thermo Scientific™ Syn-PER™ Synaptic Protein Extraction Reagent PROMO
Description
Syn-PER Reagent is used to prepare synaptosomes containing biologically active pre-and post-synaptic proteins. When used with fresh neuronal tissue or primary cultured neurons, synaptosomes prepared with the Syn-PER Reagent can be used to study synaptic transmission. The synaptosomal proteins contained in these extracts can also be used for downstream applications such as Western blotting, immunoprecipitation, enzymatic activity assays and protein-protein interaction studies. Syn-PER Synaptic Protein Extraction Reagent minimizes the degradation of phosphoproteins and is ideal for studies requiring the preservation of phosphoprotein integrity.
Highlights:
Efficient extraction – obtain up to 10μg of synaptic protein per milligram of neuronal tissue or 4μg synaptic protein per 35mm dish of primary cultured neurons (106 cells)
Gentle formulation – isolate viable synaptosomes, extract native synaptic proteins and preserve phosphoprotein integrity
Fast procedure – obtain synaptosomal suspension in less than one hour
Simple protocol – requires no ultracentrifugation steps
Recommended for:
Isolate functional synaptosomes to study neurotransmitter release; Extract pre- and post-synaptic proteins to identify changes in protein composition and function in synapses; Preserve and study labile or transient protein phosphorylation events associated with synapses
Specifications
Specifications
| Quantity | 100 mL |
| Format | Liquid |
| Product Type | Reagent |
| Volume (Metric) | 100 mL |
| Content And Storage | Upon receipt store product at 4°C. |
| Product Line | Syn-PER |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.