Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synapsin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556976 Supplier Invitrogen™ Supplier No. PA556976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDSSFMANLP NGYMTDLQRP DSSTSSPASP AMERRHPQPL AASFSSPGSS LFSSLSSAMK QAPQATSGLM EPPGPSTPIV QRPRILLVID DAHTDWSKYF HGKKVNGEIE IRVEQAEFSE LNLAAYVTGG CMVDMQVVRN Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. The protein encoded by this gene shares the synapsin family domain model, with domains A, C, and E exhibiting the highest degree of conservation. The protein contains a unique domain J, located between domains C and E. Based on this gene's localization to 22q12.3, a possible schizophrenia susceptibility locus, and the established neurobiological roles of the synapsins, this family member may represent a candidate gene for schizophrenia. The TIMP3 gene is located within an intron of this gene and is transcribed in the opposite direction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synapsin 3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYN3
Gene Accession No. O14994
Gene Alias cN28H9.2 (synapsin III); Syn3; synapsin 3; synapsin III; synapsin IIIa; synapsin-3
Gene Symbols SYN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Synapsin 3. Recombinant protein control fragment (Product #RP-91528).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.