Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin I Pan Specific Antibody, R&D Systems™

Catalog No. PPS035
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS035 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS035 Supplier R&D Systems Supplier No. PPS035
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Synapsin I Polyclonal specifically detects Synapsin I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.

Specifications

Antigen Synapsin I
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:2000 dilution, Immunoprecipitation 0.5 ug/100 ug cell lysate, Immunofluorescence 1:2000 dilution
Gene Alias SYN1, SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI
Gene Symbols SYN1
Host Species Rabbit
Immunogen Synapsin I
Purification Method Affinity Purified
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6853
Test Specificity This antibody is specific for the ∼78 kDa Synapsin I doublet in Western blots of rat brain lysates. Immunolabeling is blocked by preadsorption of the antibody with the native protein immunogen.
Reconstitution This antibody should be reconstituted in 50 μL phosphate buffered saline (137 mM NaCl, 7.5 mM Na 2 HPO 4 2.7 mM KCl, 1.5 mM KH 2 PO 4 ) before use.
Target Species Human, Mouse, Rat, Bovine
Content And Storage The lyophilized product is stable at < -20degrees C for at least one year. After reconstitution the antibody should be stored at < -20degrees C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.