Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synapsin II Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313445
This item is not returnable.
View return policy
Description
Synapsin II Polyclonal antibody specifically detects Synapsin II in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Synapsin II | |
| Polyclonal | |
| Western Blot 1:5000-1:500000, Immunohistochemistry, Immunofluorescence, Immunohistochemistry-Paraffin | |
| synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb | |
| Recombinant protein encompassing a sequence within the center region of mouse Synapsin II. The exact sequence is proprietary. | |
| 100 μg | |
| Neuroscience | |
| 6854 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction