Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Synapsin II Monoclonal Antibody (GT763)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA542341
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
Specifications
Synapsin II | |
Monoclonal | |
1 mg/mL | |
PBS with 20% glycerol and no preservative | |
Q63537, Q64332, Q92777 | |
SYN2 | |
Recombinant protein encompassing a sequence within the center region of human Synapsin II. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Western Blot, Immunocytochemistry | |
GT763 | |
Unconjugated | |
SYN2 | |
2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
Mouse | |
Protein A | |
RUO | |
20965, 29179, 6854 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction