Learn More
Synapsin II Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111854
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)SGTLKEP DSSKTP Homology: Mouse amino acid residues identical; human- 12/13 amino acid residues identical.
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
Specifications
Synapsin II | |
Polyclonal | |
Unconjugated | |
SYN2 | |
2900074L19Rik; AI836018; AI841723; SYN2; Synapsin 2; Synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20965, 29179 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q63537, Q64332 | |
SYN2 | |
Peptide corresponding to amino acid residues 438-450 of rat Synapsin-2 (Intracellular, region G) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.