Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synapsin II Rabbit anti-Human, Mouse, Rat, Clone: 7J6L0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $494.50
Specifications
Antigen | Synapsin II |
---|---|
Clone | 7J6L0 |
Dilution | Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
Synapsin II Monoclonal antibody specifically detects Synapsin II in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Synapsin II | |
Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human Synapsin II (Q92777). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7J6L0 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6854 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title