Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synapsin Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143376 Supplier Invitrogen™ Supplier No. PA5143376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Synapsins encode neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases SYN1 (Synapsin I) plays a role in regulation of axonogenesis and synaptogenesis The protein encoded serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal SYN2 (Synapsin II) encodes a neuron-specific phosphoprotein that selectively binds to small synaptic vesicles in the presynaptic nerve terminal The protein encoded by SYN3 (Synapsin III) shares the synapsin family domain model, with domains A, C, and E exhibiting the highest degree of conservation The protein contains a unique domain J, located between domains C and E.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synapsin Pan
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7
Gene Accession No. P29452, P29466, P43527, P48729, P97633, Q8BK63, Q8N752
Gene Alias SYN1a; SYN1b; synapsin I; synapsin II; synapsin III; SYNI; SYNII
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Synapsin II. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 0, 113927, 122011, 12362, 1452, 834
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.