Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptogyrin 1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306294AF532
Description
Synaptogyrin 1 Polyclonal specifically detects Synaptogyrin 1 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Synaptogyrin 1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC1939, synaptogyrin 1, synaptogyrin-1 | |
Produced in rabbits immunized with E. coli-derived Human Synaptogyrin 1 fragment. | |
0.1 mL | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
9145 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction