Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Synaptojanin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556784
Description
Immunogen sequence: HGQYSILQTA RLLPGAPQQP PKARTGISKP YNVKQIKTTN AQEAEAAIRC LLEARGGASE EALSAVAPRD LEASSEP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
The gene is a member of the inositol-polyphosphate 5-phosphatase family. The encoded protein interacts with the ras-related C3 botulinum toxin substrate 1, which causes translocation of the encoded protein to the plasma membrane where it inhibits clathrin-mediated endocytosis. Alternative splicing results in multiple transcript variants.
Specifications
Synaptojanin 2 | |
Polyclonal | |
Unconjugated | |
SYNJ2 | |
AI481647; inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; INPP5H; KIAA0348; LOW QUALITY PROTEIN: synaptojanin-2; mKIAA0348; phosphoinositide 5-phosphatase; SJ2; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; synaptojanin 2; synaptojanin-2; SYNJ2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20975, 84018, 8871 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15056, O55207, Q9D2G5 | |
SYNJ2 | |
Recombinant protein corresponding to Human Synaptojanin 2. Recombinant protein control fragment (Product #RP-94623). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction