Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptojanin 2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA530290

Catalog No. PIPA530290


Only null left
Add to Cart

Description

Description

Recommended positive controls: NT2D1. Predicted reactivity: Mouse (92%), Rat (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The gene is a member of the inositol-polyphosphate 5-phosphatase family. The encoded protein interacts with the ras-related C3 botulinum toxin substrate 1, which causes translocation of the encoded protein to the plasma membrane where it inhibits clathrin-mediated endocytosis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Synaptojanin 2
Polyclonal
Unconjugated
SYNJ2
AI481647; inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; INPP5H; KIAA0348; LOW QUALITY PROTEIN: synaptojanin-2; mKIAA0348; phosphoinositide 5-phosphatase; SJ2; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; synaptojanin 2; synaptojanin-2; SYNJ2
Rabbit
Antigen affinity chromatography
RUO
8871
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.82 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O15056
SYNJ2
Recombinant protein encompassing a sequence within the N-terminus region of human Synaptojanin 2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.