Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP225170AF405
Description
Synaptophysin Polyclonal specifically detects Synaptophysin in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Synaptophysin | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SYP | |
A synthetic peptide made to an C-terminal portion of the human Synaptophysin protein (between residues 230-313) [UniProt# P08247] | |
0.1 mL | |
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
Human, Mouse, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction