Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$378.00 - $709.50

Specifications

Antigen Synaptophysin
Concentration 1.0mg/mL
Dilution Western Blot 1:500-1:1000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:125, Immunohistochemistry-Paraffin 1:10-1:500
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300653SS
SDP
View Documents
Novus Biologicals
NB300653SS
0.025 mg
Each for $378.00
Only null left
Add to Cart
 
NB300653
SDP
View Documents
Novus Biologicals
NB300653
0.1 mg
Each for $709.50
Only null left
Add to Cart
 
Description

Description

Synaptophysin Polyclonal specifically detects Synaptophysin in Human, Mouse, Rat, Chicken, Feline, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Synaptophysin
Western Blot 1:500-1:1000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:125, Immunohistochemistry-Paraffin 1:10-1:500
Polyclonal
Rabbit
Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
SYP
IgG
Affinity Purified
The does not cross-react other synaptic vesicle proteins such as synaptotagmin, GAP45, and SNAP25 protiens.
1.0mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Chicken, Monkey
P08247
6855
Synthetic peptide taken within amino acid region 1-100 on human Synaptophysin protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
38 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.