Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Antibody (SYP/4503R), Alexa Fluor™ 350, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP308213AF350

Catalog No. NB382776


Only null left
Add to Cart

Description

Description

Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

Synaptophysin
Monoclonal
Alexa Fluor 350
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
Recombinant fragment (around aa 224-313) of human Synaptophysin (SYP) protein (exact sequence is proprietary) (Uniprot: IDP08247)
100 μg
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
6855
Store at 4°C in the dark.
IgG
ELISA, Immunohistochemistry (Paraffin)
SYP/4503R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.