Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Antibody (SYP/7976R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320528100UG
Description
Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Synaptophysin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
Recombinant fragment (around aa 274-313) of human Synaptophysin protein (exact sequence is proprietary) | |
100 μg | |
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
6855 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
SYP/7976R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction