Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Monoclonal Antibody (SYP), Abbomax

Catalog No. 502204238
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4238 Supplier Abbomax Inc Supplier No. 604310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Human pancreas Cellular location: Cytoplasmic vesicle, secretory vesicle, synaptic vesicle membrane.

Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Synaptophysin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SYP
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Syp
Gene Accession No. P08247
Gene Alias A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1
Host Species Mouse
Immunogen A recombinant protein of human Synaptophysin
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6855
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.