Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Antibody (7H12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147483AF750
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Synaptophysin | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SYP | |
Purified recombinant fragment of human Synaptophysin expressed in E. coli. [UniProtNo. P08247] | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7H12 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
Mouse | |
Ammonium sulfate precipitation | |
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
6855.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction