Learn More
Synaptophysin Mouse, anti-Human, Unconjugated, Clone: 4E12C4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Synaptophysin |
---|---|
Clone | 4E12C4 |
Concentration | 500 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9906
![]() ![]() |
Proteintech Group Inc
678641IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Synaptophysin (SYP, also known as major synaptic vesicle protein p38) is a 38-kDa integral membrane glycoprotein that regulates synaptic vesicle endocytosis. It is the most abundant synaptic vesicle protein by mass. Synaptophysin is present in neuroendocrine cells and neurons that participate in synaptic transmission. Synaptophysin is a useful marker for identification of neuroendocrine cells and neoplasms. (PMID: 3010302; 21658579)
Specifications
Synaptophysin | |
500 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat, Pig | |
Primary |
4E12C4 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse | |
6855 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.