Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Rabbit anti-Human, Clone: 2B1, Bioss

Rabbit Monoclonal Antibody
Supplier: Bioss BSM52379R

Description
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.Specifications
Synaptophysin | |
Monoclonal | |
1 μg/mL | |
TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH | |
P08247 | |
Syp | |
Recombinant human Synaptophysin between 250 to 350 amino acids. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry, Western Blot | |
2B1 | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; BOS_25912; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Rabbit | |
Protein A | |
RUO | |
6855 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction