Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Synaptophysin Recombinant Rabbit Monoclonal Antibody (RM258)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544590
Description
Synaptophysin Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
Specifications
Synaptophysin | |
Recombinant Monoclonal | |
40 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P08247 | |
Syp | |
A peptide corresponding to the C-terminus of Synaptophysin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
RM258 | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Rabbit | |
Protein A | |
RUO | |
6855 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction