Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptopodin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597078 Supplier Invitrogen™ Supplier No. PA597078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLGPHLPPPP LAPSEGRPTP CAFQIPDGSY RCLALEAEES SGEEGLQGEV GPTDLEEDEG VSRSGDDSAC RVTQGTPQLP KALGIQPPSC SREEQGASQH DDRASQDWDV VKAGQMMTAS PSPGPGPRVA QKPALGRSTS LTEKDLKEAK ARSQQIAAQL TTPPSSNSRG VQLFNRRRQR VNEFTLESHG QRGQKPSQES LRVLPSSLPG HAPGLSLSST SLPEPGPPRH PSPQSPDRGV PGHSMEGYSE EASLLRHLEK; Positive Samples: 293T, HeLa, U-87MG, Rat brain; Cellular Location: Cell junction, Cell projection, Cytoplasm, Perikaryon, cytoskeleton, dendritic spine, postsynaptic cell membrane, postsynaptic density, synapse, tight junction.

SYNPO (Synaptopodin) is an actin-associated protein in telencephalic dendrites and renal podocytes. SYNPO is tightly associated with the dendritic spine apparatus, and mice lacking the SYNPO gene lack the apparatus. These SYNPO-null mice also demonstrate an impaired ability to express long-term potentiation as well as deficits in spatial memory tasks, indicating that SYNPO is involved in the regulation of synaptic plasticity. Recent studies suggest that SYNPO is linked to neuronal calcium stores and plays a role in the calcium store-associated ability of neurons to undergo long-term plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptopodin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYNPO
Gene Accession No. Q8CC35, Q8N3V7, Q9Z327
Gene Alias 9030217H17Rik; 9130229N11; 9330140I15Rik; AW046661; KIAA1029; Synaptopodin; SYNPO
Gene Symbols SYNPO
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human SYNPO (NP_0011596801).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104027, 11346, 60324
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.