Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptoporin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563980 Supplier Invitrogen™ Supplier No. PA563980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVKVATDPKE VLLLMSACKQ PSNKCMAIHS PVMSSL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptoporin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYNPR
Gene Accession No. Q8TBG9
Gene Alias 1500003F20Rik; SPO; Synaptoporin; synaptorin; SYNPR
Gene Symbols SYNPR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Synaptoporin. Recombinant protein control fragment (Product #RP-103418).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.