Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 1/2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115994
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115994 Supplier Invitrogen™ Supplier No. PA5115994
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Synaptotagmin 1/2.

Synaptotagmins are a large gene family of synaptic vesicle type III integral membrane proteins that function as regulators of both exocytosis and endocytosis and are involved in neurotransmitter secretion from small secretory vesicles. Calcium binds to Synaptotagmin I which triggers neurotransmitter release at the synapse. Synaptotagmin II is phosphorylated by WNK1 in a process that regulates calcium-dependent interactions. Synaptotagmin III is involved in calcium-dependent exocytosis of secretory vesicles in endocrine cells and neurons. Synaptotagmin IV is expressed in neuronal tissues, and has the highest mRNA levels in the hippocampus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin 1/2
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SYT1
Gene Accession No. P21579, P21707, P29101, P46096, P46097, Q8N9I0
Gene Alias AW124717; CMS7; FLJ42519; G630098F17Rik; Inositol polyphosphate-binding protein; IP4-binding protein; mKIAA4194; MYSPC; p65; R74640; RATSYNII; SVP65; synaptotagmin 1; synaptotagmin 2; synaptotagmin I; Synaptotagmin II; synaptotagmin-1; Synaptotagmin-2; SYNII; SYT; SYT1; SYT2; SytI; sytII
Gene Symbols SYT1, SYT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SYT1(Accession P21579), corresponding to amino acid residues G176-V226.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127833, 20979, 20980, 24805, 25716, 6857
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.