Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin-1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN2180100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2180100 100 μL
ABN218025UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN2180100 Supplier MilliporeSigma Supplier No. ABN2180100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synaptotagmin-1 Polyclonal antibody specifically detects Synaptotagmin-1 in Rat samples. It is validated for Activity Assay, Immunocytochemistry, Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin-1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen BSA-conjugated linear peptide corresponding to the first 23 amino acids from the N-terminal vesicular domain of rat Synaptotagmin-1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.