Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin-11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Synaptotagmin-11 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7658
![]() ![]() |
Proteintech Group Inc
120311AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.Specifications
Synaptotagmin-11 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O08835, Q9BT88, Q9R0N3 | |
229521, 23208, 60568 | |
Synaptotagmin-11 Fusion Protein Ag2663 | |
Primary | |
-20°C | |
SYT11 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0080, Synaptotagmin 11, synaptotagmin XI, Synaptotagmin-11, SYT11, SYT12, SytXI | |
Syt11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title