Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544986 Supplier Invitrogen™ Supplier No. PA544986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AIGKIFVGSN ATGTELRHWS DMLANPRRPI AQWHSLKPEE EVDALLGKNK Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin 2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SYT2
Gene Accession No. Q8N9I0
Gene Alias CMS7; Inositol polyphosphate-binding protein; IP4-binding protein; mKIAA4194; MYSPC; R74640; RATSYNII; synaptotagmin 2; synaptotagmin I; synaptotagmin II; Synaptotagmin-2; SYNII; Syt2; SytII
Gene Symbols SYT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SYT2 (aa 370-419).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127833
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.