Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596781 Supplier Invitrogen™ Supplier No. PA596781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQRKSSKSNK TPPYKFVHVL KGVDIYPENL NSKKKFGADD KNEVKNKPAV PKNSLHLDLE KRDLNGNFPK TNLKPGSPSD LENATPKLFL EGEKESVSPE SLKSSTSLTS EEKQEKLGTL FFSLEYNFER KAFVVNIKEA RGLPAMDEQS MTS; Positive Samples: SH-SY5Y, NIH/3T3, Mouse brain, Mouse spinal cord, Mouse eye, Rat brain; Cellular Location: Cytoplasmic vesicle, Single-pass membrane protein, secretory vesicle, synaptic vesicle membrane.

May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes (PubMed:23999003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin 4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Syt4
Gene Accession No. P40749, P50232, Q9H2B2
Gene Alias HsT1192; KIAA1342; synaptotagmin 4; synaptotagmin IV; Synaptotagmin4; Synaptotagmin-4; Syt3; Syt4; SytIV
Gene Symbols Syt4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 38-190 of human SYT4 (NP_0658341).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20983, 64440, 6860
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.