Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104174 Supplier Invitrogen™ Supplier No. PA5104174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SYT4.

May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes (PubMed:23999003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin 4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Syt4
Gene Accession No. Q9H2B2
Gene Alias HsT1192; KIAA1342; synaptotagmin 4; synaptotagmin IV; Synaptotagmin4; Synaptotagmin-4; Syt3; Syt4; SytIV
Gene Symbols Syt4
Host Species Rabbit
Immunogen A synthesized peptide derived from human SYT4(Accession Q9H2B2), corresponding to amino acid residues G74-K124.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6860
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.