Learn More
Synaptotagmin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA552709
Description
Immunogen sequence: SNKTPPYKFV HVLKGVDIYP ENLNSKKKFG ADDKNEVKNK PAVPKNSLHL DLEKRDLNGN FPKTNLKPGS PSDLENATPK LFLEGEKESV SPESLKSSTS LTSEEKQEKL GTLFFSLEYN FERKAFVVNI KEARGLPAMD EQSMT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes (PubMed:23999003).
Specifications
Synaptotagmin 4 | |
Polyclonal | |
Unconjugated | |
Syt4 | |
HsT1192; KIAA1342; synaptotagmin 4; synaptotagmin IV; Synaptotagmin4; Synaptotagmin-4; Syt3; Syt4; SytIV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6860 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2B2 | |
Syt4 | |
Recombinant protein corresponding to Human Synaptotagmin 4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only