Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin 4 Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306493AF488
This item is not returnable.
View return policy
Description
Synaptotagmin 4 Polyclonal antibody specifically detects Synaptotagmin 4 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Synaptotagmin 4 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 488 | |
HsT1192, KIAA1342sytIV, synaptotagmin 4, synaptotagmin IVsynaptotagmin-4, SytIV | |
Produced in rabbits immunized with E. coli-derived Human Synaptotagmin 4 fragment. | |
0.1 mL | |
Membrane Vesicle Markers | |
6860 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction