Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Synaptotagmin 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115995
Description
Antibody detects endogenous levels of total Synaptotagmin.
Thesynaptotagmins are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis. Calcium binding tosynaptotagmin-1 participates in triggering neurotransmitter release at the synapse.
Specifications
| Synaptotagmin 5 | |
| Polyclonal | |
| Unconjugated | |
| Syt5 | |
| putative synaptotagmin V; synaptotagmin 5; synaptotagmin IX; Synaptotagmin V; synaptotagmin-5; SYT5; Syt9; SytIX; sytV | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 53420, 54309, 6861 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O00445, P47861, Q9R0N5 | |
| Syt5 | |
| A synthesized peptide derived from human SYT5(Accession O00445), corresponding to amino acid residues Q285-G335. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction