Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 7 Monoclonal Antibody (N275/14), FITC
GREENER_CHOICE

Catalog No. PIMA545589
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545589 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545589 Supplier Invitrogen™ Supplier No. MA545589
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45589 was sufficient for detection of Synaptotagmin-7 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 45kDa. Does not cross-react with Synaptotagmin-6 (or others). Can identify other isoforms bands at approximately 65kD. This antibody was formerly sold as clone S275-14.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. A similar protein in rodents mediates hormone secretion and lysosome exocytosis. In humans, expression of this gene has been associated with prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin 7
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N275/14
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene SYT7
Gene Accession No. O43581, Q62747, Q9R0N7
Gene Alias AI851541; B230112P13Rik; IPCA7; IPCA-7; PCANAP7; Prostate cancer-associated protein 7; Protein Syt7; synaptotagmin 7; synaptotagmin VII; Synaptotagmin-7; SYT7; SYT-7; SytVII; SYT-VII
Gene Symbols SYT7
Host Species Mouse
Immunogen Fusion protein amino acids 150-239 (Cytoplasmic C2A domain) of mouse Synaptotagmin-7.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54525, 59267, 9066
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.