Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114233 Supplier Invitrogen™ Supplier No. PA5114233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPSANSIIVN IIKARNLKAM DIGGTSDPYV KVWLMYKDKR VEKKKTVTMK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. A similar protein in rodents mediates hormone secretion and lysosome exocytosis. In humans, expression of this gene has been associated with prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin 7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SYT7
Gene Accession No. O43581
Gene Alias AI851541; B230112P13Rik; IPCA7; IPCA-7; PCANAP7; Prostate cancer-associated protein 7; Protein Syt7; synaptotagmin 7; synaptotagmin VII; Synaptotagmin-7; SYT7; SYT-7; SytVII; SYT-VII
Gene Symbols SYT7
Host Species Rabbit
Immunogen Immunogen sequence: NPSANSIIVN IIKARNLKAM DIGGTSDPYV KVWLMYKDKR VEKKKTVTMK (aa 277-326).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9066
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.