Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Synaptotagmin 9 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537988
Description
This antibody is predicted to react with human, rat, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
SYT9 may be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis.
Specifications
Synaptotagmin 9 | |
Polyclonal | |
Unconjugated | |
Syt9 | |
FLJ45896; synaptogamin V; synaptotagmin 5; synaptotagmin 9; synaptotagmin IX; Synaptotagmin V; Synaptotagmin9; synaptotagmin-9; SynaptotagminIX; Syt IX; Syt5; SYT9; SYT-9; SytIX; Sytv | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
60510 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9R0N9 | |
Syt9 | |
Peptide with sequence EHDSCQDFIYHLRD, from the N-terminus of SYT9 (aa 26-39). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction