Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Synaptotagmin-like Protein 4 (SYTL4), Mouse, Unlabeled, Clone: 7D2.2,

Catalog No. MABS1713MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1713MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1713MI Supplier MilliporeSigma™ Supplier No. MABS1713
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Synaptotagmin-like protein 4 (UniProt Q96C24; also known as Exophilin-2, Granuphilin) is encoded by the SYTL4 (also known as SLP4) gene (Gene ID 94121) in human. SYLT4 belongs to the synaptotagmin-like protein family, which contains an N-terminal RabBD (Rab-binding) domain and two C-terminal C2 domains. The characteristic feature of this family is the N-terminal domain, which is not found in other C-type tandem C2 proteins. SYLT4 is detected close to the plasma membrane and on secretory granules. Interacts with vesicles containing negatively charged phospholipids in a Ca2+-independent manner. It can bind to both the GTP- and GDP-bound forms of Rab27A and inhibits a specific GTP/GDP exchange cycle required for dense-core vesicle exocytosis. It is the first regulatory in the exocytosis pathway that functions by directly connecting Rab and SNARE. It modulates exocytosis of dense-core granules and secretion of hormones in the pancreas and the pituitary. It is not only essential for the docking of insulin granules, but also simultaneously imposes a fusion constraint on them through an interaction with syntaxin-1alpha fusion machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin-like Protein 4 (SYTL4)
Applications Western Blot
Classification Monoclonal
Clone 7D2.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SYTL4, SLP4
Gene Accession No. Q96C24
Host Species Mouse
Immunogen GST-tagged recombinant human synaptotagmin-like protein 4 (SYTL4) internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001123368
Test Specificity Clone 7D2.2 epitope resides within an internal region mostly missing in human spliced isoform 2. Reactivity toward isoform 2 has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.