Learn More
Synoptics™ Synbiosis Validation Kit
GREENER_CHOICESDP
Catalog No. p-7222392 Shop All Synoptics, Inc. Products
Click to view available options
For Use With (Application):
Validation of Aone Imaging
Validation of Color Count Imaging
Validation of Count Plate Imaging
Validation of East Imaging
Validation of Opka Imaging
Validation of Pour Plate Imaging
Validation of SBA Imaging
Validation of Spiral Imaging
For Use With (Equipment):
ProtoCOL
ProtoCOL and ChromaZona
ProtoCOL and Protos
Synbiosis Validation Kits are designed for use with ProtoCOL, Protos, and ChromaZona instruments and include a validation plate, a validation procedure, and a test certificate.
  • Synbiosis Validation Kits are designed for use with ProtoCOL, Protos, and ChromaZona instruments
  • Each kit includes a validation plate, a validation procedure, and a test certificate
  • Kits available for validation of: Pour Plate Imaging, Count Plate Imaging, Color Count Imaging, Aone Imagine, East Imaging, Spiral Imaging, Opka Imaging, SBA Imaging
Specifications
Product Type Validation Kit
Display Type None
Inlet Style None
Magnification None
Material Plastic, Paper
Memory None
Outlet Style None
Sample Size Single
For Use With (Application) Validation of Pour Plate Imaging
For Use With (Equipment) ProtoCOL and Protos
Includes Pour Plate Validation Plate, Pour Plate Validation Procedure, Pour Plate Test Certificate
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.