Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYNC Antibody (OTI1E7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274416V
Description
SYNC Monoclonal specifically detects SYNC in Human samples. It is validated for Western Blot.Specifications
SYNC | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SYNC | |
Full length recombinant protein of human SYNC (NP_110413) produced in SF9 cell. | |
0.1 mL | |
81493.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1E7 | |
Western Blot | |
SYNC syncoilin, intermediate filament protein, SYNC1, SYNCOILIN | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction