Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNC Monoclonal Antibody (OTI5B8)
GREENER_CHOICE

Catalog No. PIMA527186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527186 Supplier Invitrogen™ Supplier No. MA527186
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SYNC Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the intermediate filament family which contains an N-terminal head domain, followed by a central coiled-coil region and a short C-terminal tail. The protein is highly expressed in skeletal and cardiac muscle. The protein links the dystrophin associated protein complex (DAPC) to desmin filaments in muscle and may have a structural role in striated muscle. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYNC
Gene Accession No. Q9H7C4
Gene Alias 1110057H03Rik; intermediate filament protein syncoilin; SNIP4; SYNC; SYNC1; synci; SYNCOILIN; syncoilin intermediate filament 1; syncoilin, intermediate filament 1; syncoilin, intermediate filament protein; syncoilin-1
Gene Symbols SYNC
Host Species Mouse
Immunogen Full length human recombinant protein of SYNC produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81493
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.