Learn More
SYNC Mouse anti-Human, Clone: OTI5B8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809588

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the intermediate filament family which contains an N-terminal head domain, followed by a central coiled-coil region and a short C-terminal tail. The protein is highly expressed in skeletal and cardiac muscle. The protein links the dystrophin associated protein complex (DAPC) to desmin filaments in muscle and may have a structural role in striated muscle. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SYNC | |
Monoclonal | |
Unconjugated | |
SYNC | |
SYNC1, SYNCOILIN | |
Mouse | |
Affinity Chromatography | |
RUO | |
81493 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5B8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H7C4 | |
SYNC | |
Full length human recombit protein of human SYNC produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.