Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNC Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121568
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121568 20 μL
PIPA5121567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121568 Supplier Invitrogen™ Supplier No. PA5121568
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the intermediate filament family which contains an N-terminal head domain, followed by a central coiled-coil region and a short C-terminal tail. The protein is highly expressed in skeletal and cardiac muscle. The protein links the dystrophin associated protein complex (DAPC) to desmin filaments in muscle and may have a structural role in striated muscle. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNC
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SYNC
Gene Accession No. Q9EPM5, Q9H7C4
Gene Alias 1110057H03Rik; intermediate filament protein syncoilin; SNIP4; SYNC; SYNC1; synci; SYNCOILIN; syncoilin intermediate filament 1; syncoilin, intermediate filament 1; syncoilin, intermediate filament protein; syncoilin-1
Gene Symbols SYNC
Host Species Rabbit
Immunogen Ile169-Pro458 (Accession Q9H7C4), with N-terminal His and GST Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100620771, 362606, 618122, 68828, 81493
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.