Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI SyncorePlus Analyst Parallel Evaporator System
SDP

SyncorePlus Analyst is specially designed to evaporate samples to a specified volume as a pre-analytical step, with an S-Condensor the final volume is 1 mL. Includes a V-300 vacuum pump with secondary condenser, I-300 Pro interface, and F-308 recirculating chiller.

Supplier:  BUCHI 11SQA21S00121

 View more versions of this product

Catalog No. 05-405-369


Only null left
Add to Cart
Request a Quote

Description

Description

  • Safely and efficiently concentrate up to 12 samples to a predefined residual volume
  • Residual volumes range from 0.3 mL, 1 mL, to 3 mL per sample
  • Interchangeable racks and a wide range of accessories are available to create the ideal system to meet laboratory needs
  • Individual sample sealing, and the use of inert materials inhibits cross-contamination
  • A cooled appendix protects heat-sensitive analytes
Specifications

Specifications

50/60 Hz
60 to 400 rpm
115 V
SyncorePlus Analyst
Crystal Rack R-6 Analyst
520 x 500 x 325 mm
Sample Evaporation
6 x 250 mL
30 kg
20°C to 100°C
1250 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.