Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syncytin 2 Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209293
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9293 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9293 Supplier Fabgennix Inc Supplier No. SYCY2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Syncytin 2 Polyclonal antibody specifically detects Syncytin 2 in Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Syncytin 2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ERVFRD-1
Gene Alias Endogenous retrovirus group FRD member 1; endogenous retrovirus group FRD, member 1; envelope glycoprotein syncytin-A; envelope polyprotein; envFRD; ERVFRD-1; ERVFRDE1; FLJ41944; FLJ90611; GLLL6191; Gm52; HERV-FRD; HERV-FRD provirus ancestral Env polyprotein; HERV-FRD_6p24.1 provirus ancestral Env polyprotein; HERV-W/FRD; MGC87585; SU; Surface protein; Syna; syncytin 2; syncytin a; Syncytin2; syncytin-2; TM; Transmembrane protein; UNQ6191; UNQ6191/PRO20218
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 380-430 on mouse Envelope glycoprotein syncytin-B protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288602
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.