Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-1/CD138 Antibody (004), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290664AF594
Description
Syndecan-1/CD138 Monoclonal specifically detects Syndecan-1/CD138 in Mouse samples. It is validated for ELISA.Specifications
Syndecan-1/CD138 | |
Monoclonal | |
Alexa Fluor 594 | |
CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Syndecan-1/CD138 (NP_035649.1; Met1-Glu252). | |
0.1 mL | |
B Cell Development and Differentiation Markers | |
6382 | |
Mouse | |
Purified |
ELISA | |
4 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Rat SDC1 No cross-reactivity in ELISA with: Mouse SDC4 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction