Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-1/CD138 Antibody (B-A38), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314568PCP
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow CytometrySpecifications
Syndecan-1/CD138 | |
Monoclonal | |
PerCP | |
CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
U266 cell line | |
0.1 mL | |
B Cell Development and Differentiation Markers | |
6382 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
B-A38 | |
PBS | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction