Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-1/CD138 Antibody (DL-101), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143972100TESTS
Description
Syndecan-1/CD138 Monoclonal specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry.Specifications
Syndecan-1/CD138 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
Mouse | |
Affinity Purified | |
RUO | |
6382 | |
Store at 4C. Do not freeze. |
Flow Cytometry | |
DL-101 | |
FITC | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
SDC1 | |
The immunogen for this antibody was CD138. | |
100 Tests | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction