Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syndecan 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595938 Supplier Invitrogen™ Supplier No. PA595938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESRAELTSDK DMYLDNSSIE EASGVYPIDD DDYASASGSG ADEDVESPEL TTSRPLPKIL LTSAAPKVET TTLNIQNKIP AQTKSPEETD KEKVHLSDSE RKMDPAEEDT NVYTEKHSDS LFKRTE; Positive Samples: HeLa; Cellular Location: Membrane, Single-pass type I membrane protein.

Syndecans are heparan sulfate proteoglycans (HSPGs) that are involved in cell adhesion, tissue morphogenesis, differentiation, and act as low affinity co-receptors of soluble growth factors. It is involved in the mitogenic activity and signaling of Granulocyte-Macrophage Colony-stimulating factor in osteoblasts, and has been shown to have a regulatory role in matrix assembly. Syndecan-2 is expressed at epithelial-mesenchymal interfaces, and prechondrogenic and preosteogenic mesenchymal condensations, which are areas of high morphogenetic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan 2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SDC2
Gene Accession No. P34741, P43407
Gene Alias 4833414L08Rik; AA960457; CD362; cell surface-associated heparan sulfate proteoglycan 1; fibroglycan; heparan sulfate proteoglycan 1, cell surface-associated; heparan sulfate proteoglycan 1, cell surface-associated (fibroglycan); heparan sulfate proteoglycan core protein; HSPG; Hspg1; Ryudocan/syndecan 2; Sdc2; SYND2; syndecan 2; syndecan proteoglycan 2; syndecan-2
Gene Symbols SDC2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15529, 6383
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.